Gene Transcripts
- Gene Symbol
- LOC101926908
- Entrez Gene ID
- 101926908
- Gene Name
- uncharacterized LOC101926908
- Gene Aliases
- Chromosome Location
- Chr. 8: 72732045 - 72751843 on Build GRCh38
- UniGene ID
- Hs.680588
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110656.1 |
|
3 |
265 |
|
GenBank mRNA |
AK055677.1 |
- |
3 |
265 |
|
View Full Product Details