Gene Transcripts
- Gene Symbol
- LCORL
- Entrez Gene ID
- 254251
- Gene Name
- ligand dependent nuclear receptor corepressor like
- Gene Aliases
- MLR1
- Chromosome Location
- Chr. 4: 17841199 - 18023287 on Build GRCh38
- UniGene ID
- Hs.446201
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK054796.1 |
- |
1 |
621 |
|
View Full Product Details