Gene Transcripts
- Gene Symbol
- Catsperd
- Entrez Gene ID
- 106757
- Gene Name
- catsper channel auxiliary subunit delta
- Gene Aliases
- 4921529N20Rik, 4933402B14Rik, 619718, AW045609, Gm6095, Tmem146
- Chromosome Location
- Chr. 17: 56626896 - 56664460 on Build GRCm38
- UniGene ID
- Mm.23401
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK014981.1 |
- |
4 |
319 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY714503.1 |
- |
4 |
319 |
|
Chr. 17 - 56628143 - 56634147 [+] |
View Full Product Details