Gene Transcripts
- Gene Symbol
- Gm11264
- Entrez Gene ID
- 102635575
- Gene Name
- predicted gene 11264
- Gene Aliases
- OTTMUSG00000000384
- Chromosome Location
- Chr. 4: 82033309 - 82065890 on Build GRCm38
- UniGene ID
- Mm.403314
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK036601.1 |
- |
6 |
2822 |
|
View Full Product Details