Gene Transcripts
- Gene Symbol
- 2610034E01Rik
- Entrez Gene ID
- 69236
- Gene Name
- RIKEN cDNA 2610034E01 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.440848
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK011660.1 |
- |
1 |
633 |
|
AK142273.1 |
- |
1 |
431 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DV075293.1 |
- |
|
268 |
|
Chr. - - [-] |
GenBankmRNA |
BB659226.1 |
- |
1 |
431 |
|
Chr. 10 - 72658107 - 72658719 [+] |
View Full Product Details