Gene Transcripts
- Gene Symbol
- 4930434J06Rik
- Entrez Gene ID
- 74667
- Gene Name
- RIKEN cDNA 4930434J06 gene
- Gene Aliases
- Chromosome Location
- Chr. 14: 71856709 - 71920438 on Build GRCm38
- UniGene ID
- Mm.159286
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046274.1 |
|
2 |
472 |
|
GenBank mRNA |
AK015316.1 |
- |
2 |
472 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY714869.1 |
- |
2 |
472 |
|
Chr. 14 - 71856709 - 71859905 [+] |
View Full Product Details