Gene Transcripts
- Gene Symbol
- LOC100128233
- Entrez Gene ID
- 100128233
- Gene Name
- uncharacterized LOC100128233
- Gene Aliases
- Chromosome Location
- Chr. 14: 65212893 - 65222347 on Build GRCh38
- UniGene ID
- Hs.497323
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103769.1 |
|
4 |
929 |
|
GenBank mRNA |
BX161428.1 |
- |
4 |
929 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000531609.1 |
- |
4 |
929 |
|
Chr. 14 - 65212893 - 65222347 [-] |
View Full Product Details