Gene Transcripts
- Gene Symbol
- SSTR5-AS1
- Entrez Gene ID
- 146336
- Gene Name
- SSTR5 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 16: 1064081 - 1078731 on Build GRCh38
- UniGene ID
- Hs.250557
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027242.1 |
|
4 |
1646 |
|
GenBank mRNA |
AK056814.1 |
- |
4 |
1590 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000569832.5 |
- |
4 |
1588 |
|
Chr. 16 - 1064093 - 1078673 [-] |
View Full Product Details