Gene Transcripts

Gene Symbol
Klhl17
Entrez Gene ID
231003
Gene Name
kelch-like 17
Gene Aliases
AL022703, BC058738
Chromosome Location
Chr. 4: 156229044 - 156234857 on Build GRCm38
UniGene ID
Mm.423038
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198305.2 NP_938047.1 12 1872
GenBank mRNA AK049214.1 - 11 2002
AK154523.1 - 12 1810
AK170825.1 - 12 1810
AK192039.1 - 2 223
AY423764.1 - 12 1850
BC024128.1 - 1 481
BC048408.1 - 1 422
BC058738.1 - 5 459
BC139309.1 - 12 1813 8862331

View Full Product Details