Gene Transcripts

Gene Symbol
BC021891
Entrez Gene ID
234878
Gene Name
cDNA sequence BC021891
Gene Aliases
Mlk4, mKIAA1804
Chromosome Location
Chr. 8: 125910450 - 125947440 on Build GRCm38
UniGene ID
Mm.216458
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145608.2 NP_663583.2 7 1971
GenBank mRNA AK220424.1 - 7 1971
BC021891.1 - 7 1964 3156324
BC046448.1 - 4 539

View Full Product Details