Gene Transcripts

Gene Symbol
Polr2h
Entrez Gene ID
245841
Gene Name
polymerase (RNA) II (DNA directed) polypeptide H
Gene Aliases
Chromosome Location
Chr. 16: 20717826 - 20722272 on Build GRCm38
UniGene ID
Mm.288730
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145632.2 NP_663607.1 3 307
GenBank mRNA AK146429.1 - 3 332
AK161617.1 - 3 328
BC002306.1 - 3 307 3591968
BC092087.1 - 3 217 5042770
BC125612.1 - 3 291 40130127
BC125638.1 - 3 291 40130153

View Full Product Details