Gene Transcripts

Gene Symbol
Kcne2
Entrez Gene ID
246133
Gene Name
potassium voltage-gated channel, Isk-related subfamily, gene 2
Gene Aliases
2200002I16Rik, AW048273, MiRP1
Chromosome Location
Chr. 16: 92292389 - 92298137 on Build GRCm38
UniGene ID
Mm.156736
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134110.3 NP_598871.1 2 292
GenBank mRNA AK008619.1 - 2 293
BC022699.1 - 2 234 4481325

View Full Product Details