Gene Transcripts

Gene Symbol
Catsperd
Entrez Gene ID
106757
Gene Name
catsper channel auxiliary subunit delta
Gene Aliases
4921529N20Rik, 4933402B14Rik, 619718, AW045609, Gm6095, Tmem146
Chromosome Location
Chr. 17: 56626896 - 56664460 on Build GRCm38
UniGene ID
Mm.23401
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175350.3 NP_780559.2 6 448
GenBank mRNA HQ441159.1 - 6 440
HQ441160.1 - 4 222

View Full Product Details