Gene Transcripts

Gene Symbol
Mcidas
Entrez Gene ID
622408
Gene Name
multiciliate differentiation and DNA synthesis associated cell cycle protein
Gene Aliases
EG622408, Gm6320, Idas, Mci, Mcin
Chromosome Location
Chr. 13: 112993472 - 113000394 on Build GRCm38
UniGene ID
Mm.360976
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037914.3 NP_001033003.1 4 348
GenBank mRNA AK134107.1 - 4 349
BC150962.1 - 4 371 9087873

View Full Product Details