Gene Transcripts
- Gene Symbol
- Csn1s1
- Entrez Gene ID
- 24284
- Gene Name
- casein alpha s1
- Gene Aliases
- Casa, Csn1, Csna
- Chromosome Location
- Chr. 14: 21993450 - 22009308 on Build Rnor_6.0
- UniGene ID
- Rn.10744
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138874.2 |
NP_620229.2
|
12 |
421 |
|
GenBank mRNA |
J00710.1 |
- |
12 |
421 |
|
View Full Product Details