Gene Transcripts
- Gene Symbol
- Vdr
- Entrez Gene ID
- 24873
- Gene Name
- vitamin D (1,25- dihydroxyvitamin D3) receptor
- Gene Aliases
- Nr1i1
- Chromosome Location
- Chr. 7: 139344452 - 139394138 on Build Rnor_6.0
- UniGene ID
- Rn.10911
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017058.1 |
NP_058754.1
|
8 |
867 |
|
GenBank mRNA |
J04147.1 |
- |
8 |
867 |
|
View Full Product Details