Gene Transcripts

Gene Symbol
Ncr1
Entrez Gene ID
117547
Gene Name
natural cytotoxicity triggering receptor 1
Gene Aliases
KILR-1, Ly94, Nk-p46, Nkp46
Chromosome Location
Chr. 1: 73178917 - 73226504 on Build Rnor_6.0
UniGene ID
Rn.30049
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057199.1 NP_476547.1 3, 4 363
GenBank mRNA AF082533.1 - 3, 4 363
AJ012741.1 - 3, 4 353

View Full Product Details