Gene Transcripts

Gene Symbol
Fndc3b
Entrez Gene ID
294925
Gene Name
fibronectin type III domain containing 3B
Gene Aliases
RGD1311673
Chromosome Location
Chr. 2: 113109949 - 113415099 on Build Rnor_6.0
UniGene ID
Rn.757
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191704.1 NP_001178633.1 13, 14 1661
XM_006232215.2 XP_006232277.1 13, 14 1696
XM_006232216.2 XP_006232278.1 13, 14 1694

View Full Product Details