Gene Transcripts

Gene Symbol
Trem1
Entrez Gene ID
301229
Gene Name
triggering receptor expressed on myeloid cells 1
Gene Aliases
Chromosome Location
Chr. 9: 14727887 - 14743348 on Build Rnor_6.0
UniGene ID
Rn.86337
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106885.1 NP_001100355.1 3 576
XM_006244426.2 XP_006244488.1 721
XM_006244427.2 XP_006244489.1 835

View Full Product Details