Gene Transcripts

Gene Symbol
Mettl25
Entrez Gene ID
314798
Gene Name
methyltransferase like 25
Gene Aliases
RGD1310270
Chromosome Location
Chr. 7: 47847812 - 47928434 on Build Rnor_6.0
UniGene ID
Rn.212398
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108089.1 NP_001101559.1 3 560
GenBank mRNA BC127512.1 - 3 547
BC161980.1 - 3 562 9029986

View Full Product Details