Gene Transcripts

Gene Symbol
Ccdc60
Entrez Gene ID
498190
Gene Name
coiled-coil domain containing 60
Gene Aliases
Chromosome Location
Chr. 12: 46042422 - 46206748 on Build Rnor_6.0
UniGene ID
Rn.122760
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034945.1 NP_001030117.1 6 999
XM_006249464.2 XP_006249526.1 6 1016
GenBank mRNA BC103639.1 - 6 999

View Full Product Details