Gene Transcripts

Gene Symbol
METTL13
Entrez Gene ID
51603
Gene Name
methyltransferase like 13
Gene Aliases
5630401D24Rik, CGI-01, KIAA0859, feat
Chromosome Location
Chr. 1: 171781504 - 171823962 on Build GRCh38
UniGene ID
Hs.494705
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017001458.1 XP_016856947.1 1 70
GenBank mRNA BI091265.1 - 1 69

View Full Product Details