Gene Transcripts

Gene Symbol
Lats1
Entrez Gene ID
16798
Gene Name
large tumor suppressor
Gene Aliases
AW208599
Chromosome Location
Chr. 10: 7681209 - 7716461 on Build GRCm38
UniGene ID
Mm.34083
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010690.1 NP_034820.1 3 760
GenBank mRNA AK038612.1 - 3 761
BC158092.1 - 2 606 9088177
BC158123.1 - 3 755 9088196

View Full Product Details