Gene Transcripts

Gene Symbol
Cml5
Entrez Gene ID
69049
Gene Name
camello-like 5
Gene Aliases
1810018F03Rik
Chromosome Location
Chr. 6: 85817218 - 85820972 on Build GRCm38
UniGene ID
Mm.441449
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023493.2 NP_075982.2 3 797
GenBank mRNA AF187099.1 - 1 282
AK007530.1 - 3 797
BC024605.1 - 3 780 4236653

View Full Product Details