Gene Transcripts

Gene Symbol
C2CD4A
Entrez Gene ID
145741
Gene Name
C2 calcium dependent domain containing 4A
Gene Aliases
FAM148A, NLF1
Chromosome Location
Chr. 15: 62066977 - 62070917 on Build GRCh38
UniGene ID
Hs.202656
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207322.2 NP_997205.2 2 1353
GenBank mRNA AF504646.1 - 2 1327
BX648578.1 - 1 71

View Full Product Details