Gene Transcripts
- Gene Symbol
- NSUN5P2
- Entrez Gene ID
- 260294
- Gene Name
- NOP2/Sun RNA methyltransferase family member 5 pseudogene 2
- Gene Aliases
- NOL1R2, NSUN5C, WBSCR20B, WBSCR20C
- Chromosome Location
- Chr. 7: 72948293 - 72954763 on Build GRCh38
- UniGene ID
- Hs.510927
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033323.3 |
|
7, 8 |
939 |
|
GenBank mRNA |
AF416611.1 |
- |
7, 8 |
896 |
|
AF420250.1 |
- |
7, 8 |
896 |
|
AK021688.1 |
- |
7, 8 |
896 |
|
AK292107.1 |
- |
3, 4 |
372 |
|
AK303622.1 |
- |
2, 3 |
718 |
|
AK315927.1 |
- |
2, 3 |
552 |
|
AL833016.1 |
- |
6, 7 |
1582 |
|
BC007488.2 |
- |
7, 8 |
993 |
|
BC082753.1 |
- |
3, 4 |
352 |
4542435 |
BC106049.1 |
- |
3, 4 |
420 |
|
- Gene Symbol
- NSUN5
- Entrez Gene ID
- 55695
- Gene Name
- NOP2/Sun RNA methyltransferase family member 5
- Gene Aliases
- NOL1, NOL1R, NSUN5A, WBSCR20, WBSCR20A, p120, p120(NOL1)
- Chromosome Location
- Chr. 7: 73302516 - 73308867 on Build GRCh38
- UniGene ID
- Hs.510927
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001168347.2 |
NP_001161819.1
|
6, 7 |
840 |
|
NM_001168348.2 |
NP_001161820.1
|
6, 7 |
726 |
|
NM_018044.4 |
NP_060514.1
|
6, 7 |
840 |
|
NM_148956.3 |
NP_683759.1
|
6, 7 |
840 |
|
GenBank mRNA |
AF412028.1 |
- |
6, 7 |
779 |
|
AF420249.1 |
- |
6, 7 |
771 |
|
AK001129.1 |
- |
6, 7 |
779 |
|
AK098619.1 |
- |
6, 7 |
799 |
|
AK126375.1 |
- |
6, 7 |
663 |
|
AK298221.1 |
- |
6, 7 |
789 |
|
AK315361.1 |
- |
6, 7 |
781 |
|
BC008084.2 |
- |
6, 7 |
780 |
2823604 |
CN418523.1 |
- |
2, 3 |
164 |
|
- Gene Symbol
- NSUN5P1
- Entrez Gene ID
- 155400
- Gene Name
- NOP2/Sun RNA methyltransferase family member 5 pseudogene 1
- Gene Aliases
- NSUN5B, WBSCR20B
- Chromosome Location
- Chr. 7: 75410322 - 75416792 on Build GRCh38
- UniGene ID
- Hs.436034
- Species
- Homo sapiens
View Full Product Details