Gene Transcripts
- Gene Symbol
- NSUN5P2
- Entrez Gene ID
- 260294
- Gene Name
- NOP2/Sun RNA methyltransferase family member 5 pseudogene 2
- Gene Aliases
- NOL1R2, NSUN5C, WBSCR20B, WBSCR20C
- Chromosome Location
- Chr. 7: 72948293 - 72954763 on Build GRCh38
- UniGene ID
- Hs.510927
- Species
- Homo sapiens
- Gene Symbol
- NSUN5P1
- Entrez Gene ID
- 155400
- Gene Name
- NOP2/Sun RNA methyltransferase family member 5 pseudogene 1
- Gene Aliases
- NSUN5B, WBSCR20B
- Chromosome Location
- Chr. 7: 75410322 - 75416792 on Build GRCh38
- UniGene ID
- Hs.436034
- Species
- Homo sapiens
View Full Product Details