Gene Transcripts

Gene Symbol
CAPN10
Entrez Gene ID
11132
Gene Name
calpain 10
Gene Aliases
CANP10, NIDDM1
Chromosome Location
Chr. 2: 240586716 - 240599109 on Build GRCh38
UniGene ID
Hs.728234
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023083.3 NP_075571.1 12 2205
NM_023085.3 NP_075573.2 10 1740
GenBank mRNA AB058748.1 - 12 2109
AF089088.1 - 12 2186
AF089090.1 - 12 1881
AF089091.1 - 10 1585
AF089092.1 - 11 1788
AF089093.1 - 13 2100
AF089094.1 - 12 2039
AF089096.1 - 4 448
AK074807.1 - 12 2156
AK128552.1 - 9 4002
BC004260.1 - 12 2154 3606959
BC007553.1 - 12 2154 2967328

View Full Product Details