Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_080650.3 | NP_542381.1 | 7 | 706 | |
XM_011522160.1 | XP_011520462.1 | 7 | 725 | ||
XM_017022707.1 | XP_016878196.1 | 7 | 725 | ||
XM_017022708.1 | XP_016878197.1 | 7 | 725 | ||
GenBank mRNA | AK094007.1 | - | 7 | 666 | |
BC008485.2 | - | 6 | 622 | ||
BC066652.1 | - | 7 | 653 | 6066017 | |
BX537862.1 | - | 2 | 94 |