Gene Transcripts
- Gene Symbol
- Il13
- Entrez Gene ID
- 16163
- Gene Name
- interleukin 13
- Gene Aliases
- Il-13
- Chromosome Location
- Chr. 11: 53631323 - 53634702 on Build GRCm38
- UniGene ID
- Mm.1284
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008355.3 |
NP_032381.1
|
4 |
403 |
|
GenBank mRNA |
M23504.1 |
- |
4 |
404 |
|
View Full Product Details