Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010878.2 | NP_035008.2 | 3 | 820 | |
GenBank mRNA | AF043259.1 | - | 3 | 908 | |
AF084183.1 | - | 2 | 743 | ||
AK053155.1 | - | 617 | |||
AK054051.1 | - | 2 | 649 | ||
BC002015.1 | - | 3 | 820 | 3593441 |