Gene Transcripts

Gene Symbol
Nfyc
Entrez Gene ID
18046
Gene Name
nuclear transcription factor-Y gamma
Gene Aliases
Chromosome Location
Chr. 4: 120757435 - 120831579 on Build GRCm38
UniGene ID
Mm.435590
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048168.2 NP_001041633.1 3 291
NM_001277095.1 NP_001264024.1 3 364
NM_008692.4 NP_032718.2 3 364
GenBank mRNA AB006181.1 - 2 144
AK011327.1 - 3 321
AK156353.1 - 3 218
BC020117.1 - 3 290 4009737
BC053723.1 - 3 294 30053020
BC085261.1 - 3 290 6397760
CF172077.1 - 3 287
CJ092564.1 - 3 341
DV650959.1 - 3 303
U62297.1 - 2 144

View Full Product Details