Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164096.1 | NP_001157568.1 | 19 | 2127 | |
NM_025298.3 | NP_079574.2 | 20 | 2205 | ||
GenBank mRNA | AK002918.1 | - | 3 | 284 | |
AK005034.1 | - | 19 | 2128 | ||
AK012184.1 | - | 20 | 2165 | ||
AK144721.1 | - | 20 | 2170 | ||
AK146279.1 | - | 20 | 2162 | ||
BC028924.1 | - | 20 | 2126 | ||
BC037637.1 | - | 20 | 2155 | 5010681 |