Gene Transcripts
- Gene Symbol
- Polr2e
- Entrez Gene ID
- 66420
- Gene Name
- polymerase (RNA) II (DNA directed) polypeptide E
- Gene Aliases
- 2410021N14Rik, 25kDa, AW208866, RPB5, XAP4
- Chromosome Location
- Chr. 10: 80035953 - 80039659 on Build GRCm38
- UniGene ID
- Mm.18579
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025554.2 |
NP_079830.2
|
3, 4 |
450 |
|
GenBank mRNA |
AK010567.1 |
- |
3, 4 |
458 |
|
AK015823.1 |
- |
3, 4 |
448 |
|
AK088610.1 |
- |
3, 4 |
507 |
|
AK132109.1 |
- |
3, 4 |
505 |
|
AK144668.1 |
- |
3, 4 |
511 |
|
AK145201.1 |
- |
3, 4 |
514 |
|
BC005599.1 |
- |
3, 4 |
502 |
|
BC026842.1 |
- |
3, 4 |
441 |
4210827 |
BC045521.1 |
- |
3, 4 |
450 |
2647754 |
View Full Product Details