Gene Transcripts
- Gene Symbol
- Trem2
- Entrez Gene ID
- 83433
- Gene Name
- triggering receptor expressed on myeloid cells 2
- Gene Aliases
- TREM-2, Trem2a, Trem2b, Trem2c
- Chromosome Location
- Chr. 17: 48346401 - 48352276 on Build GRCm38
- UniGene ID
- Mm.261623
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF213458.1 |
- |
5 |
935 |
|
View Full Product Details