Gene Transcripts

Gene Symbol
Mrps21
Entrez Gene ID
66292
Gene Name
mitochondrial ribosomal protein S21
Gene Aliases
1810031B19Rik
Chromosome Location
Chr. 3: 95862646 - 95871604 on Build GRCm38
UniGene ID
Mm.24108
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_078479.3 NP_510964.1 2 210
GenBank mRNA AB049956.1 - 3 235
AK002588.1 - 3 238
AK003462.1 - 3 231
AK003602.1 - 3 230
AK007662.1 - 3 239
AK007838.1 - 2 221
AK008265.1 - 3 253
AK008293.1 - 3 232
AK009049.1 - 3 231
AK009703.1 - 3 239
BC027546.1 - 2 214 1515797
BC125658.1 - 2 211 40130173
BC125660.1 - 2 211 40130175

View Full Product Details