Gene Transcripts
- Gene Symbol
- NEO1
- Entrez Gene ID
- 4756
- Gene Name
- neogenin 1
- Gene Aliases
- IGDCC2, NGN, NTN1R2
- Chromosome Location
- Chr. 15: 73051715 - 73305206 on Build GRCh38
- UniGene ID
- Hs.388613
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001172623.1 |
NP_001166094.1
|
2, 3 |
635 |
|
NM_001172624.1 |
NP_001166095.1
|
2, 3 |
635 |
|
NM_002499.3 |
NP_002490.2
|
2, 3 |
635 |
|
XM_005254408.1 |
XP_005254465.1
|
3, 4 |
885 |
|
XM_011521628.2 |
XP_011519930.1
|
2, 3 |
640 |
|
XM_011521629.2 |
XP_011519931.1
|
2, 3 |
634 |
|
XM_011521630.2 |
XP_011519932.1
|
2, 3 |
634 |
|
XM_011521632.2 |
XP_011519934.1
|
2, 3 |
634 |
|
XM_011521633.2 |
XP_011519935.1
|
2, 3 |
634 |
|
XM_017022231.1 |
XP_016877720.1
|
2, 3 |
634 |
|
XM_017022232.1 |
XP_016877721.1
|
3, 4 |
885 |
|
XM_017022233.1 |
XP_016877722.1
|
2, 3 |
634 |
|
XM_017022234.1 |
XP_016877723.1
|
3, 4 |
677 |
|
XM_017022235.1 |
XP_016877724.1
|
2, 3 |
634 |
|
XM_017022236.1 |
XP_016877725.1
|
2, 3 |
634 |
|
XM_017022237.1 |
XP_016877726.1
|
2, 3 |
634 |
|
XM_017022238.1 |
XP_016877727.1
|
2, 3 |
634 |
|
GenBank mRNA |
BC117161.1 |
- |
2, 3 |
509 |
|
BC143270.1 |
- |
2, 3 |
510 |
9051772 |
BC143271.1 |
- |
2, 3 |
509 |
9051773 |
U61262.1 |
- |
2, 3 |
579 |
|
U72391.1 |
- |
2, 3 |
569 |
|
View Full Product Details