Gene Transcripts
- Gene Symbol
- CREB5
- Entrez Gene ID
- 9586
- Gene Name
- cAMP responsive element binding protein 5
- Gene Aliases
- CRE-BPA, CREB-5
- Chromosome Location
- Chr. 7: 28299321 - 28825894 on Build GRCh38
- UniGene ID
- Hs.437075
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004904.3 |
NP_004895.2
|
4, 5 |
363 |
|
NM_182898.3 |
NP_878901.2
|
4, 5 |
682 |
|
NM_182899.4 |
NP_878902.2
|
3, 4 |
338 |
|
XM_005249906.1 |
XP_005249963.1
|
4, 5 |
358 |
|
XM_017012806.1 |
XP_016868295.1
|
5, 6 |
413 |
|
XM_017012807.1 |
XP_016868296.1
|
5, 6 |
1535 |
|
XM_017012808.1 |
XP_016868297.1
|
5, 6 |
394 |
|
XM_017012809.1 |
XP_016868298.1
|
5, 6 |
419 |
|
GenBank mRNA |
AB451409.1 |
- |
3, 4 |
292 |
|
AK292916.1 |
- |
3, 4 |
351 |
|
BG199550.1 |
- |
|
289 |
|
L05515.1 |
- |
4, 5 |
682 |
|
L05911.1 |
- |
4, 5 |
364 |
|
L05913.1 |
- |
3, 4 |
338 |
|
View Full Product Details