Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127219.1 | NP_001120691.1 | 14 | 1508 | |
NM_032592.3 | NP_115981.1 | 14 | 1804 | ||
XM_005253170.3 | XP_005253227.2 | 14 | 1947 | ||
XM_005253172.2 | XP_005253229.1 | 11 | 1035 | ||
XM_006718348.2 | XP_006718411.1 | 14 | 1256 | ||
XM_011520407.1 | XP_011518709.1 | 14 | 1300 | ||
XM_011520410.2 | XP_011518712.1 | 14 | 2201 | ||
GenBank mRNA | AK057649.1 | - | 14 | 1798 | |
AL833533.1 | - | 6 | 2801 | ||
AY026508.1 | - | 14 | 1759 | ||
BC020197.1 | - | 14 | 1778 | 3632065 | |
KU178811.1 | - | 13 | 1364 | ||
KU178812.1 | - | 13 | 1364 | ||
KU178813.1 | - | 12 | 1304 |