Gene Transcripts

Gene Symbol
ATP10B
Entrez Gene ID
23120
Gene Name
ATPase phospholipid transporting 10B (putative)
Gene Aliases
ATPVB
Chromosome Location
Chr. 5: 160563120 - 160929411 on Build GRCh38
UniGene ID
Hs.109358
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011534472.2 XP_011532774.1 1 373
GenBank mRNA AK025130.1 - 1 185

View Full Product Details