Gene Transcripts
- Gene Symbol
- LNPEP
- Entrez Gene ID
- 4012
- Gene Name
- leucyl and cystinyl aminopeptidase
- Gene Aliases
- CAP, IRAP, P-LAP, PLAP
- Chromosome Location
- Chr. 5: 96935642 - 97029411 on Build GRCh38
- UniGene ID
- Hs.527199
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK096804.1 |
- |
2 |
1408 |
|
View Full Product Details