Gene Transcripts
- Gene Symbol
- SYT14P1
- Entrez Gene ID
- 401135
- Gene Name
- synaptotagmin 14 pseudogene 1
- Gene Aliases
- CHR415SYT, SYT14L, SYTDEP
- Chromosome Location
- Chr. 4: 68060610 - 68063297 on Build GRCh38
- UniGene ID
- Hs.130270
- Species
- Homo sapiens
- Gene Symbol
- SYT14
- Entrez Gene ID
- 255928
- Gene Name
- synaptotagmin 14
- Gene Aliases
- SCAR11, sytXIV
- Chromosome Location
- Chr. 1: 209938165 - 210171315 on Build GRCh38
- UniGene ID
- Hs.658866
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001146264.2 |
NP_001139736.1
|
8, 9 |
1568 |
|
NM_001256006.1 |
NP_001242935.1
|
8, 9 |
1652 |
|
NM_153262.3 |
NP_694994.2
|
7, 8 |
1447 |
|
NR_027459.2 |
|
8, 9 |
1381 |
|
XM_017000931.1 |
XP_016856420.1
|
6, 7 |
2626 |
|
GenBank mRNA |
AB102948.1 |
- |
6, 7 |
1356 |
|
AJ617624.1 |
- |
7, 8 |
1533 |
|
AJ617625.1 |
- |
8, 9 |
1295 |
|
AK091517.1 |
- |
7, 8 |
1370 |
|
BC144154.1 |
- |
8, 9 |
1501 |
|
View Full Product Details