Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320817.1 | NP_001307746.1 | 27 | 3410 | |
XM_011537482.1 | XP_011535784.1 | 28 | 3174 | ||
XM_011537483.2 | XP_011535785.1 | 28 | 3294 | ||
XM_011537484.1 | XP_011535786.1 | 27 | 3621 | ||
XM_011537485.1 | XP_011535787.1 | 27 | 3293 | ||
XM_011537486.2 | XP_011535788.2 | 27 | 3309 | ||
XM_011537487.1 | XP_011535789.1 | 27 | 3214 | ||
XM_011537488.1 | XP_011535790.1 | 27 | 3053 | ||
XM_011537490.1 | XP_011535792.1 | 27 | 3587 | ||
XM_011537491.1 | XP_011535793.1 | 25 | 3002 | ||
XM_017020494.1 | XP_016875983.1 | 27 | 3309 | ||
GenBank mRNA | BC011853.2 | - | 27 | 3370 | |
BC020208.1 | - | 27 | 3370 | 4121441 |