Gene Transcripts
- Gene Symbol
- GP5
- Entrez Gene ID
- 2814
- Gene Name
- glycoprotein V platelet
- Gene Aliases
- CD42d, GPV
- Chromosome Location
- Chr. 3: 194394821 - 194399266 on Build GRCh38
- UniGene ID
- Hs.73734
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004488.2 |
NP_004479.1
|
2 |
2163 |
|
GenBank mRNA |
L11238.1 |
- |
1 |
2204 |
|
View Full Product Details