Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130674.2 | NP_001124146.1 | 7 | 3126 | |
NM_001199671.1 | NP_001186600.1 | 8 | 3941 | ||
NM_001199672.1 | NP_001186601.1 | 8 | 3941 | ||
NM_001199673.1 | NP_001186602.1 | 6 | 2926 | ||
NM_001219.4 | NP_001210.1 | 7 | 3126 | ||
NR_074086.1 | 5 | 2700 | |||
XM_017012659.1 | XP_016868148.1 | 7 | 3491 | ||
GenBank mRNA | AF013759.1 | - | 7 | 3031 | |
HM002604.1 | - | 8 | 3942 | ||
HM002605.1 | - | 8 | 3942 | ||
HM002606.1 | - | 6 | 2868 | ||
HM002607.1 | - | 8 | 2976 | ||
HM002609.1 | - | 5 | 2784 | ||
HM002610.1 | - | 5 | 2643 | ||
HM002611.1 | - | 6 | 2926 | ||
HM002613.1 | - | 6 | 2931 | ||
HM002615.1 | - | 9 | 4136 | ||
HM002616.1 | - | 5 | 2700 |