Gene Transcripts

Gene Symbol
Mat1a
Entrez Gene ID
11720
Gene Name
methionine adenosyltransferase I, alpha
Gene Aliases
AI046368, AdoMet, Ams, MAT, MATA1, SAMS, SAMS1
Chromosome Location
Chr. 14: 41105012 - 41124428 on Build GRCm38
UniGene ID
Mm.14064
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133653.3 NP_598414.1 3 588
GenBank mRNA AK149542.1 - 2 339
AK153832.1 - 2 342
BC011211.1 - 2 326 4196928
L13622.1 - 2 340

View Full Product Details