Gene Transcripts
- Gene Symbol
- Acox1
- Entrez Gene ID
- 11430
- Gene Name
- acyl-Coenzyme A oxidase 1, palmitoyl
- Gene Aliases
- AOX, Acox, D130055E20Rik, Paox
- Chromosome Location
- Chr. 11: 116171883 - 116199045 on Build GRCm38
- UniGene ID
- Mm.356689
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF006688.1 |
- |
7 |
889 |
|
View Full Product Details