Gene Transcripts

Gene Symbol
Amph
Entrez Gene ID
218038
Gene Name
amphiphysin
Gene Aliases
Chromosome Location
Chr. 13: 18948141 - 19154032 on Build GRCm38
UniGene ID
Mm.101650
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC043718.1 - 1876
BC051425.1 - 4529
BC054718.1 - 3187 5686478

View Full Product Details