Gene Transcripts

Gene Symbol
Enox2
Entrez Gene ID
209224
Gene Name
ecto-NOX disulfide-thiol exchanger 2
Gene Aliases
APK1, Cova1, tNOX
Chromosome Location
Chr. X: 49009707 - 49288245 on Build GRCm38
UniGene ID
Mm.327730
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271447.1 NP_001258376.1 9 1365
NM_001271448.1 NP_001258377.1 8 1210
NM_001271449.1 NP_001258378.1 9 1354
NM_001271450.1 NP_001258379.1 9 1354
NM_001271451.1 NP_001258380.1 9 1354
NM_145951.5 NP_666063.1 9 1309
GenBank mRNA AK028343.1 - 9 1355
AK034058.1 - 9 1310
AK041822.1 - 9 1355
AK133956.1 - 8 1181
AK137736.1 - 9 1336
AK139897.1 - 9 1336
BC025915.1 - 9 1328 5043650

View Full Product Details