Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027578.1 | NP_081854.1 | 5 | 931 | |
NM_029018.4 | NP_083294.2 | 6 | 1150 | ||
GenBank mRNA | AK014637.1 | - | 5 | 905 | |
AK040868.1 | - | 6 | 1139 | ||
AY230199.1 | - | 6 | 1124 |